missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dibromo-3,4,5,6-tetramethylbenzene, 98%, Thermo Scientific Chemicals

Product Code. 15446387
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36321-73-0
Molecular Formula C10H12Br2
Molecular Weight (g/mol) 292.014
MDL Number MFCD00154987
InChI Key VEZJOZSIGSNYEB-UHFFFAOYSA-N
Synonym benzene,1,2-dibromo-3,4,5,6-tetramethyl, pubchem21478, benzene, 1,2-dibromo-3,4,5,6-tetramethyl, 1,2-bis bromanyl-3,4,5,6-tetramethyl-benzene
PubChem CID 2752606
IUPAC Name 1,2-dibromo-3,4,5,6-tetramethylbenzene
SMILES CC1=C(C(=C(C(=C1C)Br)Br)C)C

Specifications

Quantity 5 g
Formula Weight 292.01
Percent Purity 98%
Chemical Name or Material 1,2-Dibromo-3,4,5,6-tetramethylbenzene

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'