missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dibromoethane, 99%, Thermo Scientific Chemicals

Product Code. 11974481
Click to view available options
Quantity:
25 mL
500 mL
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic Bottle
Plastic Drum
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-93-4
Molecular Formula C2H4Br2
Molecular Weight (g/mol) 187.86
InChI Key PAAZPARNPHGIKF-UHFFFAOYSA-N
Synonym ethylene dibromide, ethylene bromide, sym-dibromoethane, ethane, 1,2-dibromo, alpha,beta-dibromoethane, bromuro di etile, 1,2-dibromaethan, 1,2-dibroomethaan, 1,2-ethylene dibromide, aadibroom
PubChem CID 7839
ChEBI CHEBI:28534
IUPAC Name 1,2-dibromoethane
SMILES C(CBr)Br

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 9.0°C to 10.0°C
Color Colorless to Yellow
Density 2.1730g/mL
Boiling Point 131.0°C to 132.0°C
Flash Point >104°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Plastic Bottle
Linear Formula BrCH2CH2Br
Refractive Index 1.5375 to 1.5395
Quantity 2.5 L
Beilstein 01,9
Fieser 01,374; 04,126; 05,187; 12,570
Merck Index 15,385
Specific Gravity 2.173
Solubility Information Solubility in water: 4g/L (20°C). Other solubilities: miscible with most organic solvents
Formula Weight 187.86
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1,2-Dibromoethane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'