missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2-dichloro-4-iodobenzene, Thermo Scientific™
Click to view available options
Quantity:
10 g
25 g
100 g
Specifications
Specifications
Assay Percent Range | 97% |
Linear Formula | IC6H3Cl2 |
Quantity | 10 g |
Formula Weight | 272.9 |
Percent Purity | 97% |
Chemical Name or Material | 1, 2-Dichloro-4-iodobenzene |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'