Learn More
1,2-Dichlorobenzene, 99+%, for spectroscopy, Thermo Scientific Chemicals
£35.58 - £134.67
Chemical Identifiers
CAS | 95-50-1 |
---|---|
Molecular Formula | C6H4Cl2 |
Molecular Weight (g/mol) | 147 |
MDL Number | MFCD00000535 |
InChI Key | RFFLAFLAYFXFSW-UHFFFAOYSA-N |
Synonym | o-dichlorobenzene, chloroben, o-dichlorbenzol, dilantin db, ortho-dichlorobenzene, 2-dichlorobenzene, o-dichlorobenzol, odcb, benzene, 1,2-dichloro, cloroben |
PubChem CID | 7239 |
ChEBI | CHEBI:35290 |
IUPAC Name | 1,2-dichlorobenzene |
SMILES | C1=CC=C(C(=C1)Cl)Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10551471
|
Thermo Scientific Acros
222055000 |
500 mL | Glass bottle |
£35.58
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10657893
|
Thermo Scientific Acros
222050025 |
2.5 L | Glass bottle |
£134.67
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for UV/VIS spectroscopyChemical Identifiers
95-50-1 | |
147 | |
RFFLAFLAYFXFSW-UHFFFAOYSA-N | |
7239 | |
1,2-dichlorobenzene |
C6H4Cl2 | |
MFCD00000535 | |
o-dichlorobenzene, chloroben, o-dichlorbenzol, dilantin db, ortho-dichlorobenzene, 2-dichlorobenzene, o-dichlorobenzol, odcb, benzene, 1,2-dichloro, cloroben | |
CHEBI:35290 | |
C1=CC=C(C(=C1)Cl)Cl |
Specifications
0.0005 meq/g max. | |
95-50-1 | |
99.0 | |
-15.0°C | |
179.0°C to 180.0°C | |
Authentic | |
C6H4Cl2 | |
1.5500 to 1.5520 | |
05, 201 | |
15, 3065 | |
o-dichlorobenzene, chloroben, o-dichlorbenzol, dilantin db, ortho-dichlorobenzene, 2-dichlorobenzene, o-dichlorobenzol, odcb, benzene, 1,2-dichloro, cloroben | |
RFFLAFLAYFXFSW-UHFFFAOYSA-N | |
1,2-dichlorobenzene | |
7239 | |
147 | |
Spectroscopy | |
1, 2-Dichlorobenzene, 99+% |
0.0003 meq/g max. | |
(1 cm cell vs water), (1 cm cell vs water) at 0nm, 0.01 at 380nm, 0.05 at 336nm, 0.3 at 300nm, 1 at 296nm | |
100.0 | |
1.3000g/mL | |
67°C | |
99% min. (GC) | |
C6H4Cl2 | |
MFCD00000535 | |
0.0005% max. | |
1.3 | |
Solubility in water: 0.13g/L(20°C). Other solubilities: miscible with most common organic solvents,miscible with alcohol,ether and benzene | |
C1=CC=C(C(=C1)Cl)Cl | |
147 | |
CHEBI:35290 | |
99+% | |
Liquid |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.
Very toxic to aquatic life with long lasting effects.
Causes skin irritation.
Harmful if inhaled.
May cause
GHS P Statement
Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritatio
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 202-425-9
missing translation for 'rtecsNumber' : CZ4500000
missing translation for 'tsca' : TSCA
- 1,2-Dichlorobenzene
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
RUO – Research Use Only