missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2-Dichloroethane, 99.5%, Extra Dry over Molecular Sieve, AcroSeal™
Alle ansehen Thermo Scientific Chemicals Produkte
Click to view available options
Quantity:
100 mL
1 L
2.5 L
Packungsgröße:
1 L
100mL
2.5 L
Beschreibung
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spezifikation
Spezifikation
| Melting Point | -35.0°C |
| Density | 1.2500g/mL |
| Boiling Point | 81.0°C to 85.0°C |
| Quantity | 100 mL |
| Flash Point | 13°C |
| Refractive Index | 1.4440 to 1.4460 |
| Specific Gravity | 1.25 |
| Linear Formula | ClCH2CH2Cl |
| Viscosity | 0.8 mPa.s (20°C) |
| Water | 0.005% max. |
| Mehr anzeigen |
Sicherheit und Handhabung
Produktidentifikator
- 1,2-Dichloroethane
- Danger
- Flammable liquid Category 2
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Acute toxicity Category 3
- Specific target organ toxicity Category 3
- Carcinogenicity Category 1B
- H225-Highly flammable liquid and vapour.
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H331-Toxic if inhaled.
- H335-May cause respiratory irritation.
- H336-May cause drowsiness or dizziness.
- H350-May cause cancer.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P311-Call a POISON CENTER/doctor/ .
- P362+P364-Take off contaminated clothing and wash it before reuse.
RUO – Research Use Only
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur