missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2-Dichloroethane, 99+%, ACS reagent, meets the requirements of Reag. Ph.Eur.
Alle ansehen Thermo Scientific Chemicals Produkte
missing translation for 'orderingAttributeHoverText'
Quantity:
500 mL
1 L
2.5 L
Packaging:
Glass Bottle
missing translation for 'unitSize'
1 L
2.5 L
500mL
Beschreibung
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spezifikation
Spezifikation
| Melting Point | -35.0°C |
| CAS Min % | 99.0 |
| CAS Max % | 100.0 |
| Density | 1.2500g/mL |
| Boiling Point | 81.0°C to 85.0°C |
| Quantity | 500 mL |
| Flash Point | 13°C |
| Packaging | Glass Bottle |
| Physical Form | Liquid |
| Refractive Index | 1.4440 to 1.4460 |
| Mehr anzeigen |
Sicherheit und Handhabung
Produktidentifikator
- 1,2-Dichloroethane
- Danger
- Acute toxicity Category 3
- Acute toxicity Category 4
- Carcinogenicity Category 1B
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H225-Highly flammable liquid and vapour.
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H331-Toxic if inhaled.
- H335-May cause respiratory irritation.
- H336-May cause drowsiness or dizziness.
- H350-May cause cancer.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P311-Call a POISON CENTER/doctor/ .
- MIXTURE LIST-Contains : 1,2-Dichloroethane
RUO – Research Use Only
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur