missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dichloroethane, 99+%, for spectroscopy ACS, Thermo Scientific Chemicals

Product Code. 10571804
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
Glass Bottle
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For UV/VIS spectroscopy
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-06-2
Molecular Formula C2H4Cl2
Molecular Weight (g/mol) 98.95
MDL Number MFCD00000963
InChI Key WSLDOOZREJYCGB-UHFFFAOYSA-N
Synonym ethylene dichloride, ethylene chloride, ethane, 1,2-dichloro, dutch liquid, dutch oil, ethane dichloride, glycol dichloride, aethylenchlorid, dichloro-1,2-ethane, dichloremulsion
PubChem CID 11
ChEBI CHEBI:27789
IUPAC Name 1,2-dichloroethane
SMILES ClCCCl

Specifications

Melting Point -35.0°C
Absorbance 0.01 max. at 255nm, 0.01 max. at 400nm, 0.02 max. at 250nm, 0.05 max. at 245nm, 0.1 max. at 240nm, 0.2 max. at 235nm, 0.5 max. at 230nm, 1 max. at 228nm
CAS Min % 99.0
CAS Max % 100.0
Density 1.2500g/mL
Boiling Point 81.0°C to 85.0°C
Quantity 2.5 L
Flash Point 13°C
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.4440 to 1.4460
Assay Percent Range 99+%
Residue after Evaporation 0.002% max.
Specific Gravity 1.25
Titratable Acid 0.0003 meq/g max.
Linear Formula ClCH2CH2Cl
Vapor Pressure Vapor Pressure: 65mmHg at 29°C
Viscosity 0.8 mPa.s (20°C)
Beilstein 01, 84
Water 0.03% max. (Coulometric)
Merck Index 15, 3851
Solubility Information Solubility in water: 8.7g/L (20°C). Other solubilities: miscible with most common organic solvents
Formula Weight 98.96
Percent Purity ≥99%
Grade ACS
Chemical Name or Material 1,2-Dichloroethane, 99+%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'