Learn More
1,2-Dichloroethane, 99+%, Thermo Scientific Chemicals
Description
1,2-Dichloroethane is used to make vinyl chloride and chlorinated solvents (trichloroethane, trichloroethylene, perchloroethylene, and vinylidene) to remove grease, resins, glue and dirt. It is used as a solvent in the manufacture of polystyrene and SBR latex. It is added to leaded petrol as an anti-knock compound. It is also used in polyvinyl chloride (PVC) pipes, packaging materials, furniture, auto mobile parts, wall coverings and housewares. It is also used generally as an intermediate for other organic chemical compounds and as a solvent. It is a useful solvent for the isomer distribution in the acetylation of benzene and toluene under the influence of aluminum chloride.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1,2-Dichloroethane is used to make vinyl chloride and chlorinated solvents (trichloroethane, trichloroethylene, perchloroethylene, and vinylidene) to remove grease, resins, glue and dirt. It is used as a solvent in the manufacture of polystyrene and SBR latex. It is added to leaded petrol as an anti-knock compound. It is also used in polyvinyl chloride (PVC) pipes, packaging materials, furniture, auto mobile parts, wall coverings and housewares. It is also used generally as an intermediate for other organic chemical compounds and as a solvent. It is a useful solvent for the isomer distribution in the acetylation of benzene and toluene under the influence of aluminum chloride.
Solubility
Miscible with methanol, diethyl ether, n-octanol and acetone. Very slightly miscible with water.
Notes
Incompatible with strong oxidizing agents, strong reducing agents, bases and alkali metals.
Specifications
Melting Point | -35°C |
Density | 1.256 |
Boiling Point | 83°C |
Quantity | 500 mL |
Flash Point | 15°C (59°F) |
Refractive Index | 1.4448 |
Linear Formula | ClCH2CH2Cl |
UN Number | UN1184 |
Beilstein | 605264 |
Merck Index | 14,3797 |
Show More |
Safety and Handling
- 1,2-Dichloroethane
- Danger
- Acute toxicity Category 3
- Acute toxicity Category 4
- Carcinogenicity Category 1B
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H225-Highly flammable liquid and vapour.
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H331-Toxic if inhaled.
- H335-May cause respiratory irritation.
- H336-May cause drowsiness or dizziness.
- H350-May cause cancer.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P311-Call a POISON CENTER/doctor/ .
- P362-Take off contaminated clothing.
- MIXTURE LIST-Contains : ethylene dichloride
RUO – Research Use Only