missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2-Diiodotetrafluoroethane, 96%, stab. with copper, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
1,2-Diiodotetrafluoroethane is used as an organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,2-Diiodotetrafluoroethane is used as an organic chemical synthesis intermediate.
Notes
Light sensitive. Incompatible with oxidizing agents.
1,2-Diiodotetrafluoroethane is used as an organic chemical synthesis intermediate.
Notes
Light sensitive. Incompatible with oxidizing agents.
Specifications
Specifications
Melting Point | -21°C |
Density | 2.629 |
Boiling Point | 112°C to 113°C |
Flash Point | None |
Refractive Index | 1.495 |
Quantity | 1 g |
Beilstein | 1740350 |
Sensitivity | Light sensitive |
Formula Weight | 353.82 |
Percent Purity | 96% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'