missing translation for 'onlineSavingsMsg'
Learn More
1,2-Dimethoxyethane, ≥99%, Reagent Grade, Inhibitor-free, Honeywell™ Product Code.: 15657840

1,2-Dimethoxyethane, ≥99%, Reagent Grade, Inhibitor-free, Honeywell™

Product Code. 15657840 Shop All Honeywell Research Chemicals Products
1 L, Glass bottle
Click to view available options
Quantity:
100 mL
500 mL
1 L
2.5 L
20 L
Packaging:
Glass bottle
Stainless steel can
This item is not returnable. View return policy

Product Code. 15657840

Brand: Honeywell Research Chemicals E274081L

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Reagent Grade, inhibitor-free,≥99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-71-4
Molecular Formula C4H10O2
Molecular Weight (g/mol) 90.122
MDL Number MFCD00008502
InChI Key XTHFKEDIFFGKHM-UHFFFAOYSA-N
Synonym monoglyme, ethylene glycol dimethyl ether, glyme, dimethyl cellosolve, egdme, ethane, 1,2-dimethoxy, dimethoxyethane, 2,5-dioxahexane, glycol dimethyl ether, ethylene dimethyl ether
PubChem CID 8071
ChEBI CHEBI:42263
IUPAC Name 1,2-dimethoxyethane
SMILES COCCOC

Specifications

Melting Point -58°C
Color Colorless
Density 0.867g/mL at 25°C
pH Neutral
Boiling Point 85°C (1013 hPa)
Flash Point −2°C (28.4°F)
Packaging Glass bottle
Linear Formula CH3OCH2CH2OCH3
Quantity 1 L
UN Number UN2252
Beilstein 1209237
Vapor Density 3.1 (20°C, vs air)
Formula Weight 90.12g/mol
Percent Purity ≥99%
Chemical Name or Material 1,2-Dimethoxyethane
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 1,2-Dimethoxyethane
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 2
  • Reproductive toxicity Category 1B
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H332-Harmful if inhaled.
  • H360FD-May damage fertility. May damage the unborn child.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 1,2-dimethoxyethane
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1,2-Dimethoxyethane, ≥99%, Reagent Grade, Inhibitor-free, Honeywell™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.