missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dimethyl-3-hydroxy-4-pyridone, 99+%, Thermo Scientific Chemicals

Product Code. 10702491
Click to view available options
Quantity:
5 g
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30652-11-0
Molecular Formula C7H9NO2
Molecular Weight (g/mol) 139.15
MDL Number MFCD00134497
InChI Key TZXKOCQBRNJULO-UHFFFAOYSA-N
Synonym deferiprone, ferriprox, 3-hydroxy-1,2-dimethyl-4 1h-pyridone, 3-hydroxy-1,2-dimethylpyridin-4 1h-one, 1,2-dimethyl-3-hydroxy-4-pyridone, 1,2-dimethyl-3-hydroxypyrid-4-one, 1,2-dimethyl-3-hydroxypyridine-4-one, cp20 chelating agent, unii-2bty8kh53l, ferriprox tn
PubChem CID 2972
ChEBI CHEBI:68554
IUPAC Name 3-hydroxy-1,2-dimethylpyridin-4-one
SMILES CC1=C(C(=O)C=CN1C)O

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 271°C to 273°C
Color White
pH 6 to 8 (1% soln.)
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Quantity 10 g
Merck Index 15, 2862
Solubility Information Solubility in water: soluble in hot water. Other solubilities: soluble in dmso
Formula Weight 139.15
Percent Purity 99+%
Physical Form Crystalline Powder or Needles
Chemical Name or Material 1, 2-Dimethyl-3-hydroxy-4-pyridone
Show More Show Less
compliance-icons
Product Identifier
  • 1,2-Dimethyl-3-hydroxy-4-pyridone
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'