Learn More
1,2-Distearoyl-sn-glycero-3-phosphorylethanolamine, MedChemExpress
MedChemExpress 1,2-Distearoyl-sn-glycero-3-phosphorylethanolamine (DSPE) is a phosphoethanolamine (PE) lipid that can be used in the synthesis of liposomes.
Brand: Medchem Express HY-112530
This item is not returnable.
View return policy
Description
1,2-Distearoyl-sn-glycero-3-phosphorylethanolamine (DSPE) is a phosphoethanolamine (PE) lipid that can be used in the synthesis of liposomes.Specifications
1,2-Distearoyl-sn-glycero-3-phosphorylethanolamine | |
White | |
C41H82NO8P | |
DSPE | |
CCCCCCCCCCCCCCCCCC(O[C@@H](COP(OCCN)(O)=O)COC(CCCCCCCCCCCCCCCCC)=O)=O | |
748.07 | |
Research | |
Metabolism-sugar/lipid metabolism |
1069-79-0 | |
Research | |
100 mg | |
H2O : 1 mg/mL (1.34 mM; Need ultrasonic) ∣Ethanol : < 1 mg/mL (ultrasonic;adjust pH to 5 with HCl) (insoluble) ∣DMSO : < 1 mg/mL (ultrasonic) (insoluble or slightly soluble) | |
748.07 | |
97.0% | |
Solid |
Research purposes only