missing translation for 'onlineSavingsMsg'
Learn More
1,2-Epoxyhexane, 97%, Thermo Scientific Chemicals Product Code.: 10187803

1,2-Epoxyhexane, 97%, Thermo Scientific Chemicals

Product Code. 10187803
100 mL, Glass bottle
Click to view available options
Quantity:
5 mL
25 mL
100 mL
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10187803

Brand: Thermo Scientific Acros 335551000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1436-34-6
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.16
MDL Number MFCD00005154
InChI Key WHNBDXQTMPYBAT-UHFFFAOYSA-N
Synonym 1,2-epoxyhexane, butyloxirane, oxirane, butyl, 1-hexene oxide, 1-hexene epoxide, 1,2-hexene oxide, epoxy-n-hexane, hexylene oxide, hexane, 1,2-epoxy, hexene, monooxideShow MoreShow Less
PubChem CID 15036
IUPAC Name 2-butyloxirane
SMILES CCCCC1CO1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless
Density 0.8300g/mL
Boiling Point 118°C to 120°C
Flash Point 15°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Refractive Index 1.4040 to 1.407
Quantity 100 mL
Specific Gravity 0.83
Formula Weight 100.16
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1, 2-Epoxyhexane
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0436462 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number335550250,335550050,335551000
Lot NumberA0412201 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number335550250,335550050,335551000
Lot NumberA0447623 Certificate TypeCertificate of Analysis Date16/11/2022 Catalog Number335550250,335550050,335551000
Lot NumberA0386059 Certificate TypeCertificate of Analysis Date08/07/2020 Catalog Number335550250,335550050,335551000

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1,2-Epoxyhexane, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.