Learn More
1,2-Epoxyhexane, 97%, Thermo Scientific Chemicals
£10.81 - £94.80
Chemical Identifiers
CAS | 1436-34-6 |
---|---|
Molecular Formula | C6H12O |
Molecular Weight (g/mol) | 100.16 |
MDL Number | MFCD00005154 |
InChI Key | WHNBDXQTMPYBAT-UHFFFAOYSA-N |
Synonym | 1,2-epoxyhexane, butyloxirane, oxirane, butyl, 1-hexene oxide, 1-hexene epoxide, 1,2-hexene oxide, epoxy-n-hexane, hexylene oxide, hexane, 1,2-epoxy, hexene, monooxide |
PubChem CID | 15036 |
IUPAC Name | 2-butyloxirane |
SMILES | CCCCC1CO1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10192351
|
Thermo Scientific Acros
335550050 |
5 mL | Glass bottle | ||||||
10419062
|
Thermo Scientific Acros
335550250 |
25 mL | Glass bottle |
£32.20
25mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10187803
|
Thermo Scientific Acros
335551000 |
100 mL | Glass bottle |
£94.80
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1436-34-6 | |
100.16 | |
WHNBDXQTMPYBAT-UHFFFAOYSA-N | |
15036 | |
CCCCC1CO1 |
C6H12O | |
MFCD00005154 | |
1,2-epoxyhexane, butyloxirane, oxirane, butyl, 1-hexene oxide, 1-hexene epoxide, 1,2-hexene oxide, epoxy-n-hexane, hexylene oxide, hexane, 1,2-epoxy, hexene, monooxide | |
2-butyloxirane |
Specifications
1436-34-6 | |
100.0 | |
0.8300g/mL | |
15°C | |
97% | |
1.4040 to 1.407 | |
0.83 | |
WHNBDXQTMPYBAT-UHFFFAOYSA-N | |
2-butyloxirane | |
15036 | |
97% | |
1, 2-Epoxyhexane |
96.0 | |
Colorless | |
118°C to 120°C | |
Authentic | |
C6H12O | |
MFCD00005154 | |
1,2-epoxyhexane, butyloxirane, oxirane, butyl, 1-hexene oxide, 1-hexene epoxide, 1,2-hexene oxide, epoxy-n-hexane, hexylene oxide, hexane, 1,2-epoxy, hexene, monooxide | |
CCCCC1CO1 | |
100.16 | |
100.16 | |
Liquid |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Highly flammable liquid and vapor.
May cause respiratory irritation.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 215-864-6
RUO – Research Use Only