missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Fluoro-4-nitrophenyl)-4-methylpiperazine, 97%, Thermo Scientific™

Product Code. 15480619
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 221198-29-4
Molecular Formula C11H14FN3O2
Molecular Weight (g/mol) 239.25
MDL Number MFCD02633484
InChI Key UWZIHNDVFUJRHD-UHFFFAOYSA-N
Synonym 1-2-fluoro-4-nitrophenyl-4-methylpiperazine, piperazine, 1-2-fluoro-4-nitrophenyl-4-methyl, acmc-209fse, cambridge id 5846150, 1-2-fluoro-4-nitro-phenyl-4-methyl-piperazine, fluoronitrophenylmethylpiperazine, 4-2-fluoro-4-nitrophenyl-1-methylpiperazine
PubChem CID 2783054
IUPAC Name 1-(2-fluoro-4-nitrophenyl)-4-methylpiperazine
SMILES CN1CCN(CC1)C1=C(F)C=C(C=C1)[N+]([O-])=O

Specifications

Quantity 1 g
Formula Weight 239.25
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 1-(2-Fluoro-4-nitrophenyl)-4-methylpiperazine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'