Learn More
1-(2-Fluorophenyl)piperazine, 97%, Thermo Scientific Chemicals
£195.00
Chemical Identifiers
CAS | 1011-15-0 |
---|---|
Molecular Formula | C10H13FN2 |
Molecular Weight (g/mol) | 180.22 |
MDL Number | MFCD00040726 |
InChI Key | IVTZRJKKXSKXKO-UHFFFAOYSA-N |
Synonym | 1-2-fluorophenyl piperazine, 1-2-fluorophenyl-piperazine, 1-2-fluoro-phenyl-piperazine, piperazine, 1-2-fluorophenyl, chembl9429, 2-fluorophenyl piperazine, ofpp, pubchem8573, 1-2-fluorophenyl piperazinehydrochloride, 2-fluorophenylpiperazine |
PubChem CID | 70529 |
IUPAC Name | 1-(2-fluorophenyl)piperazine |
SMILES | C1CN(CCN1)C2=CC=CC=C2F |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10511121
|
Thermo Scientific Acros
268691000 |
100 g | Plastic bottle | ||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1011-15-0 | |
180.22 | |
IVTZRJKKXSKXKO-UHFFFAOYSA-N | |
70529 | |
C1CN(CCN1)C2=CC=CC=C2F |
C10H13FN2 | |
MFCD00040726 | |
1-2-fluorophenyl piperazine, 1-2-fluorophenyl-piperazine, 1-2-fluoro-phenyl-piperazine, piperazine, 1-2-fluorophenyl, chembl9429, 2-fluorophenyl piperazine, ofpp, pubchem8573, 1-2-fluorophenyl piperazinehydrochloride, 2-fluorophenylpiperazine | |
1-(2-fluorophenyl)piperazine |
Specifications
1011-15-0 | |
100.0 | |
1.1410g/mL | |
96% min. (GC) | |
1.5550 to 1.558 | |
1.141 | |
IVTZRJKKXSKXKO-UHFFFAOYSA-N | |
1-(2-fluorophenyl)piperazine | |
70529 | |
97% | |
1-(2-Fluorophenyl)piperazine, 97% |
96.0 | |
Colorless to Yellow | |
Authentic | |
C10H13FN2 | |
MFCD00040726 | |
1-2-fluorophenyl piperazine, 1-2-fluorophenyl-piperazine, 1-2-fluoro-phenyl-piperazine, piperazine, 1-2-fluorophenyl, chembl9429, 2-fluorophenyl piperazine, ofpp, pubchem8573, 1-2-fluorophenyl piperazinehydrochloride, 2-fluorophenylpiperazine | |
C1CN(CCN1)C2=CC=CC=C2F | |
180.22 | |
180.22 | |
Liquid |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 213-780-4
missing translation for 'tsca' : TSCA
- 1-(2-Fluorophenyl)piperazine
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 C
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H312-Harmful in contact with skin.
- H302-Harmful if swallowed.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only