missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Fluorophenyl)pyrrole, 98%, Thermo Scientific™

Product Code. 15426067
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89096-77-5
Molecular Formula C10H8FN
Molecular Weight (g/mol) 161.18
MDL Number MFCD02665231
InChI Key LMLYMAWSPZKOFR-UHFFFAOYSA-N
Synonym 1-2-fluorophenyl-1h-pyrrole, 1-2-fluorophenyl pyrrole, 1h-pyrrole, 1-2-fluorophenyl, n-2-fluorophenyl pyrrole, acmc-20lhmh, 1-2-fluoro-phenyl-1h-pyrrole
PubChem CID 2767601
SMILES FC1=CC=CC=C1N1C=CC=C1

Specifications

Boiling Point 64°C (1.1 mmHg)
Refractive Index 1.5105
Quantity 25 g
IUPAC Name 1-(2-fluorophenyl)-1H-pyrrole
Formula Weight 161.18
Percent Purity 98%
Chemical Name or Material 1-(2-Fluorophenyl)pyrrole
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.