missing translation for 'onlineSavingsMsg'
Learn More

1-(2-morpholinoethyl)-1h-pyrazole-4-boronic acid, 97%, Thermo Scientific™

Product Code. 10451124
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

CAS 864754-18-7
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C15H26BN3O3
MDL Number MFCD08062342
Quantity 1 g
Synonym 4-2-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazol-1-yl ethyl morpholine, 1-2-morpholinoethyl-1h-pyrazole-4-boronic acid pinacol ester, 1-2-morpholinoethyl-1h-pyrazole-4-boronic acid, pinacol ester, 1-2-morpholinoethyl pyrazole-4-boronic acid pinacol ester, 1-2-morpholinoethyl-1h-pyrazol-4-boronic acid pinacol ester, 4-2-4-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazol-1-yl ethyl morpholine, 4-2-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazol-1-yl ethyl morpholine
InChI Key FBAPTUAEBQMVEY-UHFFFAOYSA-N
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CN(N=C2)CCN3CCOCC3
IUPAC Name 4-[2-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazol-1-yl]ethyl]morpholine
Molecular Weight (g/mol) 307.201
PubChem CID 17749924
Formula Weight 307.2
Percent Purity 97%
Chemical Name or Material 1-(2-Morpholinoethyl)-1H-pyrazole-4-boronic acid, pinacol ester
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'