missing translation for 'onlineSavingsMsg'
Learn More

1,2-Octanediol, 98+%, Thermo Scientific Chemicals

Product Code. 10206690
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1117-86-8
Molecular Formula C8H18O2
Molecular Weight (g/mol) 146.23
MDL Number MFCD00010738
InChI Key AEIJTFQOBWATKX-UHFFFAOYSA-N
Synonym 1,2-octanediol, 1,2-dihydroxyoctane, 1,2-octylene glycol, capyryl glycol, racemic 1,2-octanediol, acmc-2099as, dsstox_cid_16646, dsstox_rid_79289
PubChem CID 14231
ChEBI CHEBI:34056
IUPAC Name octane-1,2-diol
SMILES CCCCCCC(CO)O

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point 28°C to 31°C
Color Colorless to White
Density 0.9140g/mL
Boiling Point 267°C
Flash Point 138°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)5CH(OH)CH2OH
Quantity 50 g
Beilstein 01, III, 2217
Specific Gravity 0.914
Solubility Information Solubility in water: 3g/L (20°C).
Viscosity 35.5 mPa.s (40°C)
Formula Weight 146.23
Percent Purity 98+%
Physical Form Low Melting Solid
Chemical Name or Material 1, 2-Octanediol, 98+ %
Show More Show Less
compliance-icons
Product Identifier
  • 1,2-Octanediol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'