missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(2-Phenoxyphenyl)ethanone, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
Specifications
Specifications
| CAS Min % | 96.5 |
| CAS Max % | 100.0 |
| Assay Percent Range | 97% |
| Packaging | Amber glass bottle |
| Quantity | 250 mg |
| Formula Weight | 212.25 |
| Percent Purity | 97% |
| Chemical Name or Material | 1-(2-Phenoxyphenyl)ethanone |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
1-(2-Phenoxyphenyl)ethanone, 97%, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction

