missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(2-Phenyl-1,3-thiazol-4-yl)ethan-1-one, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
Specifications
CAS Min % | 95.0 |
Assay Percent Range | 95% |
Packaging | Amber Glass Bottle |
Quantity | 1 g |
Formula Weight | 203.26 |
Percent Purity | 95% |
Chemical Name or Material | 1-(2-Phenyl-1, 3-thiazol-4-yl)ethan-1-one |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'