missing translation for 'onlineSavingsMsg'
Learn More

1,2-Propanediol, 99+%, for analysis, Thermo Scientific Chemicals

Product Code. 10022910
Click to view available options
Quantity:
25 mL
250 mL
1 L
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57-55-6
Molecular Formula C3H8O2
Molecular Weight (g/mol) 76.09
MDL Number MFCD00064272
InChI Key DNIAPMSPPWPWGF-UHFFFAOYSA-N
Synonym 1,2-propanediol, propylene glycol, 1,2-dihydroxypropane, 2-hydroxypropanol, 1,2-propylene glycol, methylethyl glycol, methylethylene glycol, monopropylene glycol, isopropylene glycol, dowfrost
PubChem CID 1030
ChEBI CHEBI:16997
IUPAC Name propane-1,2-diol
SMILES CC(CO)O

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point -60.0°C
Density 1.0300g/mL
Boiling Point 187.0°C
Flash Point 99°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula CH3CH(OH)CH2OH
Refractive Index 1.4310 to 1.4330
Quantity 25 mL
Beilstein 01, 472
Merck Index 15, 7968
Specific Gravity 1.03
Solubility Information Solubility in water: completely soluble. Other solubilities: soluble in ether, miscible with acetone and chloroform
Viscosity 45 mPa.s (20°C)
Formula Weight 76.09
Percent Purity 99+%
Grade Analysis
Physical Form Viscous Liquid
Chemical Name or Material 1, 2-Propanediol
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.