missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Pyridyl)piperazine, 99%, Thermo Scientific Chemicals

Product Code. 11307455
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

1-(2-Pyridyl)piperazine may be employed as reagent for the determination of both aliphatic and aromatic isocyanates in air by reversed-phase HPLC. It may be employed as reagent for the fluorometric determination of airborne diisocyantes. It is a compound that belongs to a class of selective α2-adrenoceptor antagonists. 1-(2-Pyridyl)piperazine shows sympatholytic activity. 1-(2-Pyridyl)piperazine is also a metabolite of Azaperone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-(2-Pyridyl)piperazine may be employed as reagent for the determination of both aliphatic and aromatic isocyanates in air by reversed-phase HPLC. It may be employed as reagent for the fluorometric determination of airborne diisocyantes. It is a compound that belongs to a class of selective α2-adrenoceptor antagonists. 1-(2-Pyridyl)piperazine shows sympatholytic activity. 1-(2-Pyridyl)piperazine is also a metabolite of Azaperone.

Solubility
Soluble in Chloroform, Methanol.

Notes
Air Sensitive. Store away from air and oxidizing agents. Incompatible with carbondioxide, Strong acids, Acid chlorides, Acid anhydrides.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34803-66-2
Molecular Formula C9H13N3
Molecular Weight (g/mol) 163.224
MDL Number MFCD00006216
InChI Key GZRKXKUVVPSREJ-UHFFFAOYSA-N
Synonym 1-2-pyridyl piperazine, 1-pyridin-2-yl piperazine, 1-pyridin-2-yl-piperazine, 1-2-pyridinyl piperazine, piperazine, 1-2-pyridinyl, 2-pyridylpiperazine, pyridinylpiperazine, 2-1-piperazinyl pyridine
PubChem CID 94459
IUPAC Name 1-pyridin-2-ylpiperazine
SMILES C1CN(CCN1)C2=CC=CC=N2

Specifications

Density 1.072
Boiling Point 120°C to 122°C (2 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.595
Quantity 5 g
UN Number UN2735
Beilstein 140423
Sensitivity Air Sensitive
Solubility Information Soluble in Chloroform,Methanol.
Formula Weight 163.22
Percent Purity 99%
Chemical Name or Material 1-(2-Pyridyl)piperazine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'