Learn More
1-(2-Pyrimidinyl)homopiperazine, Thermo Scientific Chemicals
£69.82 - £295.00
Chemical Identifiers
CAS | 21279-57-2 |
---|---|
Molecular Formula | C9H14N4 |
Molecular Weight (g/mol) | 178.24 |
MDL Number | MFCD06738258 |
InChI Key | LZOGPVCTSDAYIP-UHFFFAOYSA-N |
Synonym | 1-pyrimidin-2-yl-1,4-diazepane, 1-2-pyrimidinyl homopiperazine, 1-pyrimidin-2-yl-1,4 diazepane, 1-pyrimidin-2-yl-1,4-diazaperhydroepine, acmc-20aorx, 1-2-pyrimidyl homopiperazine, 1-2-pyrimidyl-homopiperazine |
PubChem CID | 2772409 |
IUPAC Name | 1-pyrimidin-2-yl-1,4-diazepane |
SMILES | C1CNCCN(C1)C1=NC=CC=N1 |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
21279-57-2 | |
178.24 | |
LZOGPVCTSDAYIP-UHFFFAOYSA-N | |
2772409 | |
C1CNCCN(C1)C1=NC=CC=N1 |
C9H14N4 | |
MFCD06738258 | |
1-pyrimidin-2-yl-1,4-diazepane, 1-2-pyrimidinyl homopiperazine, 1-pyrimidin-2-yl-1,4 diazepane, 1-pyrimidin-2-yl-1,4-diazaperhydroepine, acmc-20aorx, 1-2-pyrimidyl homopiperazine, 1-2-pyrimidyl-homopiperazine | |
1-pyrimidin-2-yl-1,4-diazepane |
Specifications
21279-57-2 | |
MFCD06738258 | |
Air Sensitive | |
LZOGPVCTSDAYIP-UHFFFAOYSA-N | |
1-pyrimidin-2-yl-1,4-diazepane | |
2772409 | |
1-(2-Pyrimidinyl)homopiperazine |
C9H14N4 | |
UN2735 | |
1-pyrimidin-2-yl-1,4-diazepane, 1-2-pyrimidinyl homopiperazine, 1-pyrimidin-2-yl-1,4 diazepane, 1-pyrimidin-2-yl-1,4-diazaperhydroepine, acmc-20aorx, 1-2-pyrimidyl homopiperazine, 1-2-pyrimidyl-homopiperazine | |
C1CNCCN(C1)C1=NC=CC=N1 | |
178.24 | |
178.24 |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only