missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Thiazolyl)piperazine hydrochloride, 97+%, Thermo Scientific™

Product Code. 15489047
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 209733-13-1
Molecular Formula C7H12ClN3S
Molecular Weight (g/mol) 205.704
MDL Number MFCD08752687
InChI Key BIHQYKGSYPXHRN-UHFFFAOYSA-N
Synonym 2-piperazin-1-yl thiazole hydrochloride, 1-2-thiazolyl piperazine hydrochloride, 1-2-thiazolyl-piperazine hydrochloride, piperazine, 1-2-thiazolyl-hydrochloride, 2-piperazin-1-ylthiazole hydrochloride, 2-piperazinyl-1,3-thiazole, chloride, 2-1-piperazinyl thiazole hydrochloride, 2-piperazin-1-yl thiazole hcl, 2-piperazin-1-yl-1,3-thiazole hydrochloride, 1-1,3-thiazol-2-yl piperazine-hydrogen chloride 1/1
PubChem CID 23467106
IUPAC Name 2-piperazin-1-yl-1,3-thiazole;hydrochloride
SMILES C1CN(CCN1)C2=NC=CS2.Cl

Specifications

Quantity 1 g
Formula Weight 205.71
Percent Purity ≥97%
Chemical Name or Material 1-(2-Thiazolyl)piperazine hydrochloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'