missing translation for 'onlineSavingsMsg'
Learn More
1,2-Vinylenebis(triphenylphosphonium bromide) monohydrate, 98%, Thermo Scientific™ Product Code.: 11364384

1,2-Vinylenebis(triphenylphosphonium bromide) monohydrate, 98%, Thermo Scientific™

Product Code. 11364384
50 g
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy

Product Code. 11364384

Brand: Thermo Scientific Alfa Aesar L00956.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54770-27-3
Molecular Formula C38H32Br2P2
Molecular Weight (g/mol) 710.43
MDL Number MFCD00040548
InChI Key TVFYBHGFLHYJNV-GLDAUVFXSA-L
Synonym triphenyl e-2-triphenylphosphonio ethenyl phosphonium dibromide
PubChem CID 123134366
SMILES [Br-].[Br-].C(=C/[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)\[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point ∼236°C to 250°C
Quantity 50 g
Beilstein 3583793
Sensitivity Hygroscopic
IUPAC Name triphenyl[(1E)-2-(triphenylphosphaniumyl)ethenyl]phosphanium dibromide
Formula Weight 728.45 (710.44 Anhydrous)
Percent Purity 98%
Chemical Name or Material 1,2-Vinylenebis(triphenylphosphonium bromide) monohydrate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10136203 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Lot Number10136806 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1,2-Vinylenebis(triphenylphosphonium bromide) monohydrate, 98%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.