missing translation for 'onlineSavingsMsg'
Learn More

1-[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-1H-pyrazole, 97%, Thermo Scientific™

Product Code. 10423403
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 852227-94-2
Molecular Formula C15H19BN2O2
Molecular Weight (g/mol) 270.14
MDL Number MFCD07368520
InChI Key GVSCNAOZDQCWJJ-UHFFFAOYSA-N
Synonym 1-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl-1h-pyrazole, 1-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl pyrazole, 1-3-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl pyrazole, 3-pyrazol-1-yl-phenylboronic acid pinacol ester, 3-1h-pyrazol-1-yl benzeneboronic acid, pinacol ester, 3-1h-pyrazol-1-yl phenyl boronic acid pinacol ester, 4,4,5,5-tetramethyl-2-3-pyrazolylphenyl-1,3,2-dioxaborolane, 1-3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl phenyl-1h-pyrazole, 1-3-4,4,5,5-tetramethyl-1,3,2-dioxaboranlan-2-yl phenyl-1h-pyrazole
PubChem CID 4961249
IUPAC Name 1-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]pyrazole
SMILES CC1(C)OB(OC1(C)C)C1=CC(=CC=C1)N1C=CC=N1

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 270.14
Percent Purity 97%
Chemical Name or Material 1-[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-1H-pyrazole
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.