missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-[3-(4-Methylpiperazin-1-yl)phenyl]methanamine, 97%, Thermo Scientific™
£49.23 - £234.00
Chemical Identifiers
CAS | 672325-37-0 |
---|---|
Molecular Formula | C12H19N3 |
Molecular Weight (g/mol) | 205.305 |
InChI Key | VKTLTRIKVYJVOX-UHFFFAOYSA-N |
Synonym | 1-3-4-methylpiperazin-1-yl phenyl methanamine, 3-4-methyl-1-piperazinyl benzenemethanamine, 3-4-methylpiperazin-1-yl phenyl methanamine, 3-4-methyl-1-piperazinyl-benzenemethanamine, 3-4-methyl-piperazin-1-yl-benzylamine, 3-4-methyl-1-piperazinyl-benzene methanamine, 3-4-methylpiperazinyl phenyl methylamine, pubchem12998 |
PubChem CID | 4961252 |
IUPAC Name | [3-(4-methylpiperazin-1-yl)phenyl]methanamine |
SMILES | CN1CCN(CC1)C2=CC=CC(=C2)CN |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10351053
|
Thermo Scientific Maybridge
CC35913CB |
250 mg | Amber Glass Bottle | N/A | |||||
10695713
|
Thermo Scientific Maybridge
CC35913DA |
1 g | Amber Glass Bottle | N/A | |||||
10193324
|
Thermo Scientific Maybridge
CC35913DE |
5 g | Amber Glass Bottle | N/A | |||||
Description
Chemical Identifiers
672325-37-0 | |
205.305 | |
1-3-4-methylpiperazin-1-yl phenyl methanamine, 3-4-methyl-1-piperazinyl benzenemethanamine, 3-4-methylpiperazin-1-yl phenyl methanamine, 3-4-methyl-1-piperazinyl-benzenemethanamine, 3-4-methyl-piperazin-1-yl-benzylamine, 3-4-methyl-1-piperazinyl-benzene methanamine, 3-4-methylpiperazinyl phenyl methylamine, pubchem12998 | |
[3-(4-methylpiperazin-1-yl)phenyl]methanamine |
C12H19N3 | |
VKTLTRIKVYJVOX-UHFFFAOYSA-N | |
4961252 | |
CN1CCN(CC1)C2=CC=CC(=C2)CN |
Specifications
672325-37-0 | |
C12H19N3 | |
VKTLTRIKVYJVOX-UHFFFAOYSA-N | |
[3-(4-methylpiperazin-1-yl)phenyl]methanamine | |
4961252 | |
97% |
97% | |
1-3-4-methylpiperazin-1-yl phenyl methanamine, 3-4-methyl-1-piperazinyl benzenemethanamine, 3-4-methylpiperazin-1-yl phenyl methanamine, 3-4-methyl-1-piperazinyl-benzenemethanamine, 3-4-methyl-piperazin-1-yl-benzylamine, 3-4-methyl-1-piperazinyl-benzene methanamine, 3-4-methylpiperazinyl phenyl methylamine, pubchem12998 | |
CN1CCN(CC1)C2=CC=CC(=C2)CN | |
205.305 | |
205.3 | |
1-[3-(4-Methylpiperazin-1-yl)phenyl]methanamine |