missing translation for 'onlineSavingsMsg'
Learn More

1-(3,5-Dichlorophenyl)-2-hydroxyethylamine, 94%, Thermo Scientific™

Product Code. 10364402
Click to view available options
Quantity:
500 mg
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 372144-00-8
Assay Percent Range 93% min. (HPLC)
Packaging Glass bottle
Molecular Formula C8H9Cl2NO
Quantity 500 mg
Synonym 2-amino-2-3,5-dichlorophenyl ethanol, 1-3,5-dichlorophenyl-2-hydroxyethylamine, 2-amino-2-3,5-dichlorophenyl ethan-1-ol, benzeneethanol, b-amino-3,5-dichloro
InChI Key LRORZXIRKBZZAG-UHFFFAOYSA-N
SMILES C1=C(C=C(C=C1Cl)Cl)C(CO)N
IUPAC Name 2-amino-2-(3,5-dichlorophenyl)ethanol
Molecular Weight (g/mol) 206.07
PubChem CID 2734585
Formula Weight 206.07
Percent Purity 94%
Chemical Name or Material 1-(3, 5-Dichlorophenyl)-2-hydroxyethylamine
Show More Show Less
compliance-icons
Product Identifier
  • 1-(3,5-Dichlorophenyl)-2-hydroxyethylamine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'