missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Tri-n-propylhexahydro-1,3,5-triazine, 96%, Thermo Scientific™

Product Code. 15488686
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13036-81-2
Molecular Formula C12H27N3
Molecular Weight (g/mol) 213.37
MDL Number MFCD00040461
InChI Key DPHBVWJGMBYPMK-UHFFFAOYSA-N
Synonym 1,3,5-triazine, hexahydro-1,3,5-tripropyl, hexahydro-1,3,5-tripropyl-s-triazine, s-triazine, hexahydro-1,3,5-tripropyl, hexahydro-1,3,5-tripropyl-1,3,5-triazine, #, 1,3,5-tri-n-propyl hexahydro-s-triazine, 1,3,5-tripropylhexahydro-1,3,5-triazine, 1,3,5-tri-n-propylhexahydro-1,3,5-triazine
PubChem CID 83068
IUPAC Name 1,3,5-tripropyl-1,3,5-triazinane
SMILES CCCN1CN(CCC)CN(CCC)C1

Specifications

Color Colorless
Density 0.880 g/mL
Boiling Point 94°C (0.7 mmHg)
Refractive Index 1.466
Quantity 250 mg
Beilstein 111423
Formula Weight 213.37
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 1,3,5-Tri-n-propylhexahydro-1,3,5-triazine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'