missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Triformylbenzene, 98%, Thermo Scientific Chemicals

Product Code. 11974781
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3163-76-6
Molecular Formula C9H6O3
Molecular Weight (g/mol) 162.14
MDL Number MFCD00182470
InChI Key AEKQNAANFVOBCU-UHFFFAOYSA-N
Synonym 1,3,5-benzenetricarboxaldehyde, 1,3,5-triformylbenzene, trimesaldehyde, 1,3,5-benzenetricarbaldehyde, benzene-1,3,5-tricarboxaldehyde, trimesic formaldehyde, 1,3,5-triformyl benzene, bzoh31, aekqnaanfvobcu-uhfffaoysa
PubChem CID 2747968
IUPAC Name benzene-1,3,5-tricarbaldehyde
SMILES O=CC1=CC(C=O)=CC(C=O)=C1

Specifications

Melting Point 156.0°C to 158.0°C
Assay Percent Range 98%
Quantity 5 g
Formula Weight 162.14
Percent Purity 98%
Chemical Name or Material 1, 3, 5-Triformylbenzene
compliance-icons
Product Identifier
  • 1,3,5-Triformylbenzene
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'