missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Trimethoxybenzene, 99%

Product Code. 11461738
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 11461738

Brand: Thermo Scientific Alfa Aesar A13981.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 621-23-8 | C9H12O3 | 168.19 g/mol

1,3,5-Trimethoxybenzene is a key component of the Chinese rose odor. It is used as a cosmetic agents. It was employed as secondary standard in quantitative proton NMR spectroscopy of pharmaceuticals. It cleaves p-methoxybenzyl protecting group on various alcohols and acids

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3,5-Trimethoxybenzene is a key component of the Chinese rose odor. It is used as a cosmetic agents. It was employed as secondary standard in quantitative proton NMR spectroscopy of pharmaceuticals. It cleaves p-methoxybenzyl protecting group on various alcohols and acids

Solubility
Insoluble in water.

Notes
Store in a cool, dry, well-ventilated area. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 621-23-8
Molecular Formula C9H12O3
Molecular Weight (g/mol) 168.19
MDL Number MFCD00008385
InChI Key LKUDPHPHKOZXCD-UHFFFAOYSA-N
Synonym phloroglucinol trimethyl ether, sym-trimethoxybenzene, benzene, 1,3,5-trimethoxy, 1,3,5-trimethyoxybenzene, unii-00vji3vg3d, 1,3,5-trimethoxy benzene, o,o,o-1,3,5-trimethylresorcinol, 00vji3vg3d, dsstox_cid_25963, dsstox_rid_81259
PubChem CID 69301
ChEBI CHEBI:31038
IUPAC Name 1,3,5-trimethoxybenzene
SMILES COC1=CC(OC)=CC(OC)=C1

Specifications

Melting Point 51°C to 53°C
Boiling Point 255°C
Flash Point 85°C (185°F)
Quantity 50 g
Beilstein 1307993
Solubility Information Insoluble in water.
Formula Weight 168.19
Percent Purity 99%
Chemical Name or Material 1,3,5-Trimethoxybenzene
compliance-icons
Product Identifier
  • 1,3,5-Trimethoxybenzene
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : 1,3,5-Trimethoxybenzene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.