missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Trimethyl-1,3,5-tris(3,3,3-trifluoropropyl)cyclotrisiloxane, 97%, Thermo Scientific Chemicals

Product Code. 11329325
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2374-14-3
Molecular Formula C12H21F9O3Si3
Molecular Weight (g/mol) 468.538
MDL Number MFCD00054667
InChI Key URZHQOCYXDNFGN-UHFFFAOYSA-N
Synonym silicone fluid, fluorosilicone trimer, 1,3,5-trimethyl-1,3,5-tris 3,3,3-trifluoropropyl cyclotrisiloxane, 1,3,5-tris 3,3,3-trifluoropropyl methyl cyclotrisiloxane, cyclotrisiloxane, 2,4,6-trimethyl-2,4,6-tris 3,3,3-trifluoropropyl, fluorosilicone cyclic trimer, b071 cut ii, 3,3,3-trifluoropropyl methylcyclotrisiloxane, 2,4,6-trimethyl-2,4,6-tris 3,3,3-trifluoropropyl cyclotrisiloxane, 3,3,3-trifluoropropyl methylsiloxane cyclic trimer
PubChem CID 75410
IUPAC Name 2,4,6-trimethyl-2,4,6-tris(3,3,3-trifluoropropyl)-1,3,5,2,4,6-trioxatrisilinane
SMILES C[Si]1(O[Si](O[Si](O1)(C)CCC(F)(F)F)(C)CCC(F)(F)F)CCC(F)(F)F

Specifications

Boiling Point 135°C (22mmHg)
Melting Point 34°C to 36°C
Quantity 25 g
UN Number UN2811
Beilstein 1809096
Formula Weight 468.54
Percent Purity 97%
Density 1.24
Chemical Name or Material 1,3,5-Trimethyl-1,3,5-tris(3,3,3-trifluoropropyl)cyclotrisiloxane

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'