missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Trimethyl-1H-pyrazole-4-carbaldehyde, Thermo Scientific™

Product Code. 10101022
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 2644-93-1
Molecular Formula C7H10N2O
Molecular Weight (g/mol) 138.17
InChI Key HMTUBXVXHHITGO-UHFFFAOYSA-N
Synonym 1,3,5-trimethyl-1h-pyrazole-4-carbaldehyde, 1,3,5-trimethyl-1h-pyrazole-4-carboxaldehyde, 1h-pyrazole-4-carboxaldehyde, 1,3,5-trimethyl, n-methyl-3,5-dimethyl-4-formylpyrazole, 1,3,5-trimethyl-1h-pyrazole-4-carbaldehyde?, akos pao-0265, timtec-bb sbb000012, acmc-1cq3n, art-chem-bb b006543, ksc496q8h
PubChem CID 736515
IUPAC Name 1,3,5-trimethylpyrazole-4-carbaldehyde
SMILES CC1=C(C(=NN1C)C)C=O

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 138.17
Percent Purity 97%
Chemical Name or Material 1, 3, 5-Trimethyl-1H-pyrazole-4-carbaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'