missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3,5-Trimethyl-1H-pyrazole-4-carboxylic acid, 97%, Thermo Scientific™
£27.60 - £535.00
Chemical Identifiers
CAS | 1125-29-7 |
---|---|
Molecular Formula | C7H10N2O2 |
Molecular Weight (g/mol) | 154.17 |
MDL Number | MFCD00159648 |
InChI Key | NOIOGQJFLIPRBI-UHFFFAOYSA-N |
Synonym | 1,3,5-trimethyl-1h-pyrazole-4-carboxylic acid, trimethyl-1h-pyrazole-4-carboxylic acid, trimethylpyrazole-4-carboxylic acid, 1,3,5-trimethyl-4-pyrazolecarboxylic acid, 1h-pyrazole-4-carboxylic acid, 1,3,5-trimethyl, akos pao-0337, rarechem al be 0403, art-chem-bb b006363, cambridge id 5264152, 1,3,5-trimethyl-1h-pyrazol-4-carboxylic acid |
PubChem CID | 736514 |
IUPAC Name | 1,3,5-trimethylpyrazole-4-carboxylic acid |
SMILES | CN1N=C(C)C(C(O)=O)=C1C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10436462
|
Thermo Scientific Maybridge
CC14701CB |
250 mg | Amber Glass Bottle |
£27.60
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10262682
|
Thermo Scientific Maybridge
CC14701DA |
1 g | Amber Glass Bottle |
£95.55
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10716295
|
Thermo Scientific Maybridge
CC14701EA |
10 g | Amber Glass Bottle |
£535.00
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
1125-29-7 | |
154.17 | |
NOIOGQJFLIPRBI-UHFFFAOYSA-N | |
736514 | |
CN1N=C(C)C(C(O)=O)=C1C |
C7H10N2O2 | |
MFCD00159648 | |
1,3,5-trimethyl-1h-pyrazole-4-carboxylic acid, trimethyl-1h-pyrazole-4-carboxylic acid, trimethylpyrazole-4-carboxylic acid, 1,3,5-trimethyl-4-pyrazolecarboxylic acid, 1h-pyrazole-4-carboxylic acid, 1,3,5-trimethyl, akos pao-0337, rarechem al be 0403, art-chem-bb b006363, cambridge id 5264152, 1,3,5-trimethyl-1h-pyrazol-4-carboxylic acid | |
1,3,5-trimethylpyrazole-4-carboxylic acid |
Specifications
1125-29-7 | |
C7H10N2O2 | |
1,3,5-trimethyl-1h-pyrazole-4-carboxylic acid, trimethyl-1h-pyrazole-4-carboxylic acid, trimethylpyrazole-4-carboxylic acid, 1,3,5-trimethyl-4-pyrazolecarboxylic acid, 1h-pyrazole-4-carboxylic acid, 1,3,5-trimethyl, akos pao-0337, rarechem al be 0403, art-chem-bb b006363, cambridge id 5264152, 1,3,5-trimethyl-1h-pyrazol-4-carboxylic acid | |
CN1N=C(C)C(C(O)=O)=C1C | |
154.17 | |
154.17 | |
1, 3, 5-Trimethyl-1H-pyrazole-4-carboxylic acid |
97% | |
MFCD00159648 | |
NOIOGQJFLIPRBI-UHFFFAOYSA-N | |
1,3,5-trimethylpyrazole-4-carboxylic acid | |
736514 | |
97% |
Safety and Handling
Warning