missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Tris(chloromethyl)-2,4,6-trimethylbenzene, 97%, Thermo Scientific Chemicals

Product Code. 15470087
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3849-01-2
Molecular Formula C12H15Cl3
Molecular Weight (g/mol) 265.60
MDL Number MFCD00229016
InChI Key PHQFMPNZCIHSPC-UHFFFAOYSA-N
IUPAC Name 1,3,5-tris(chloromethyl)-2,4,6-trimethylbenzene

Specifications

Color White
Quantity 250 mg
UN Number UN3261
Beilstein 2369523
Synonym 1,3,5-tris chloromethyl-2,4,6-trimethylbenzene, 1,3,5-tris-chloromethyl-2,4,6-trimethyl-benzene, benzene, 1,3,5-tris chloromethyl-2,4,6-trimethyl, ksc224a3n, tris-chloromethyl-mesitylene, 2,4,6-tris chloromethyl mesitylene, tris chloromethyl-2,4,6-trimethylbenzene, 1,3,5-trimethyl-2,4,6-tris chloromethyl benzene, benzene,3,5-tris chloromethyl-2,4,6-trimethyl, 1,3,5-tris chloromethyl-2,4,6-trimethyl-benzene
SMILES CC1=C(C(=C(C(=C1CCl)C)CCl)C)CCl
PubChem CID 291188
Formula Weight 265.6
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 1,3,5-Tris(chloromethyl)-2,4,6-trimethylbenzene
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'