missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-[3-Amino-5-(4-chlorophenyl)-2-thienyl]ethan-1-one, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
10 g
Specifications
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Packaging | Amber glass bottle |
Quantity | 250 mg |
IUPAC Name | 1-[3-amino-5-(4-chlorophenyl)thiophen-2-yl]ethan-1-one |
Formula Weight | 251.74 |
Percent Purity | 97% |
Chemical Name or Material | 1-[3-Amino-5-(4-chlorophenyl)-2-thienyl]ethan-1-one |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'