missing translation for 'onlineSavingsMsg'
Learn More

1,3-Benzenediboronic acid, 97%, Thermo Scientific™

Product Code. 11401377
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
1,3-benzenediboronic acid is used to prepare 1,3-didydrxybenzene by oxidation reaction using alkaline hydrogen peroxide.

Solubility
Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4612-28-6
Molecular Formula C6H8B2O4
Molecular Weight (g/mol) 165.746
MDL Number MFCD03092915
InChI Key UXPAASVRXBULRG-UHFFFAOYSA-N
Synonym 1,3-benzenediboronic acid, 1,3-phenylenediboronic acid, 1,3-benzendiboronic acid, 1,3-benzenediboronicacid, m-phenylenediboronic acid, 3-dihydroxyboranyl phenyl boronic acid, 3-dihydroxyboranyl phenylboronic acid, m-benzenediboronic acid, pubchem17415, 1,3-benzendibororic acid
PubChem CID 230477
IUPAC Name (3-boronophenyl)boronic acid
SMILES B(C1=CC(=CC=C1)B(O)O)(O)O

Specifications

Melting Point >300°C
Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 165.75
Percent Purity 97%
Chemical Name or Material 1,3-Benzenediboronic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'