missing translation for 'onlineSavingsMsg'
Learn More

1,3-Benzenedithiol, 97%, Thermo Scientific Chemicals

Product Code. 11358736
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

1,3-Benzenedithiol is used to improve power conversion efficiency in cadmium selenide nanorods/poly(3-hexylthiophene) hybrid solar cells. It acts as an intermediate as well as involved in the preparation of 1,3-bis(-styrylthio)-benzol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Benzenedithiol is used to improve power conversion efficiency in cadmium selenide nanorods/poly(3-hexylthiophene) hybrid solar cells. It acts as an intermediate as well as involved in the preparation of 1,3-bis(-styrylthio)-benzol.

Solubility
Miscible with acetonitrile.

Notes
Incompatible with strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 626-04-0
Molecular Formula C6H6S2
Molecular Weight (g/mol) 142.23
MDL Number MFCD00004842
InChI Key ZWOASCVFHSYHOB-UHFFFAOYSA-N
Synonym 1,3-benzenedithiol, 1,3-dimercaptobenzene, dithioresorcin, dithioresorcinol, 1,3-benzedithiol, acmc-20ap5g, ksc357g0j
PubChem CID 522062
IUPAC Name benzene-1,3-dithiol
SMILES SC1=CC(S)=CC=C1

Specifications

Density 1.24
Boiling Point 244°C
Flash Point >112°C (234°F)
Odor Stench
Refractive Index 1.6625
Quantity 5 g
Beilstein 1634412
Sensitivity Air sensitive
Solubility Information Miscible with acetonitrile.
Formula Weight 142.24
Percent Purity 97%
Chemical Name or Material 1,3-Benzenedithiol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'