missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Benzodioxol-5-ylacetyl chloride, ≥97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
10 g
Specifications
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Quantity | 10 g |
IUPAC Name | 2-(2H-1,3-benzodioxol-5-yl)acetyl chloride |
Formula Weight | 198.6 |
Percent Purity | 97% |
Chemical Name or Material | 1, 3-Benzodioxol-5-ylacetyl chloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'