missing translation for 'onlineSavingsMsg'
Learn More
1,3-Benzodioxole, 99%, Thermo Scientific Chemicals Product Code.: 11422267

1,3-Benzodioxole, 99%, Thermo Scientific Chemicals

Product Code. 11422267
50 g
Click to view available options
Quantity:
50 g
250 g
1000 g
This item is not returnable. View return policy

Product Code. 11422267

Brand: Thermo Scientific Alfa Aesar A14192.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

1,3-Benzodioxole is useful in gemological stimulant detection. It is also used as a precursor for perfumes, photo initiators, agrochemicals and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Benzodioxole is useful in gemological stimulant detection. It is also used as a precursor for perfumes, photo initiators, agrochemicals and pharmaceuticals.

Solubility
Soluble in water (2g/L).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 274-09-9
Molecular Formula C7H6O2
Molecular Weight (g/mol) 122.123
MDL Number MFCD00005818
InChI Key FTNJQNQLEGKTGD-UHFFFAOYSA-N
Synonym 1,2-methylenedioxybenzene, benzodioxole, benzo d 1,3 dioxole, 1,3-dioxaindan, 3,4-methylenedioxybenzene, 2h-1,3-benzodioxole, 1,2-methylenedioxy benzene, 1,3-dioxindan, methylenedioxybenzene, o-methylenedioxy benzene
PubChem CID 9229
ChEBI CHEBI:38732
IUPAC Name 1,3-benzodioxole
SMILES C1OC2=CC=CC=C2O1

Specifications

Melting Point -18°C
Density 1.185
Boiling Point 172°C to 173°C
Flash Point 55°C (131°F)
Odor Aromatic
Refractive Index 1.539
Quantity 50 g
Beilstein 115506
Solubility Information Soluble in water (2g/L).
Formula Weight 122.12
Percent Purity 99%
Chemical Name or Material 1,3-Benzodioxole
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10253322 Certificate TypeCertificate of Analysis Date04/12/2024 Catalog NumberA14192.30,A14192.18,A14192.0B
Lot Number10252259 Certificate TypeCertificate of Analysis Date02/09/2024 Catalog NumberA14192.30,A14192.18,A14192.0B
Lot Number10251005 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberA14192.30,A14192.18,A14192.0B
Lot Number10243214 Certificate TypeCertificate of Analysis Date05/09/2022 Catalog NumberA14192.30,A14192.18,A14192.0B
Lot Number10144368 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14192.30,A14192.18,A14192.0B

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1,3-Benzodioxole, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.