missing translation for 'onlineSavingsMsg'
Learn More

1,3-bis(2,4,6-trimethylphenyl)imidazolidinium chloride, 97%

Product Code. 10669604 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10669604

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 173035-10-4 | C21H27ClN2 | 342.91 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 173035-10-4
Molecular Formula C21H27ClN2
Molecular Weight (g/mol) 342.91
MDL Number MFCD09039279
InChI Key COGMCBFILULEOS-UHFFFAOYSA-M
Synonym 1,3-dimesitylimidazolidine hydrochloride, 1,3-bis 2,4,6-trimethylphenyl-imidazolidinium-chloride, 1,3-dimesitylimidazolidine, chloride salt, 1,3-bis-2,4,6-trimethyl-phenyl-imidazolidin-1-ium chloride, 1,3-bis 2,4,6-trimethylphenyl imidazolidine hydrochloride
IUPAC Name 1,3-bis(2,4,6-trimethylphenyl)-4,5-dihydro-3H-1λ⁵-imidazol-1-ylium chloride
SMILES [Cl-].CC1=CC(C)=C(N2CC[N+](=C2)C2=C(C)C=C(C)C=C2C)C(C)=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 280°C to 286°C
Color Beige to Brown-Pink
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 342.91
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 1,3-Bis(2,4,6-trimethylphenyl)imidazolidinium chloride
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.