missing translation for 'onlineSavingsMsg'
Learn More

1,3-Bis(dichloromethyl)-1,1,3,3-tetramethyldisiloxane, 94%, Thermo Scientific™

£17.90 - £123.00

Chemical Identifiers

CAS 2943-70-6
Molecular Formula C6H14Cl4OSi2
Molecular Weight (g/mol) 300.147
MDL Number MFCD00040856
InChI Key YMDCHYWOAHMASL-UHFFFAOYSA-N
PubChem CID 76260
IUPAC Name dichloromethyl-[dichloromethyl(dimethyl)silyl]oxy-dimethylsilane
SMILES C[Si](C)(C(Cl)Cl)O[Si](C)(C)C(Cl)Cl
View More Specs

Products 2
Produktcode Marke Quantity Preis Menge & Verfügbarkeit  
Produktcode Marke Quantity Preis Menge & Verfügbarkeit  
11337615
missing translation for 'viewDocuments'
Thermo Scientific Alfa Aesar
L05858.06
5 g
£17.90
5g
Please to purchase this item. Need a web account? Register with us today!
11357615
missing translation for 'viewDocuments'
Thermo Scientific Alfa Aesar
L05858.22
100 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Beschreibung

Beschreibung

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Spezifikation

Chemische Identifikatoren

2943-70-6
300.147
YMDCHYWOAHMASL-UHFFFAOYSA-N
dichloromethyl-[dichloromethyl(dimethyl)silyl]oxy-dimethylsilane
C6H14Cl4OSi2
MFCD00040856
76260
C[Si](C)(C(Cl)Cl)O[Si](C)(C)C(Cl)Cl

Spezifikation

149°C to 150°C (40mmHg)
C6H14Cl4OSi2
1762720
1,3-bis dichloromethyl-1,1,3,3-tetramethyldisiloxane, 1,3-bis dichloromethyl tetramethyldisiloxane, bis dichloromethyl tetramethyldisiloxane, disiloxane, 1,3-bis dichloromethyl-1,1,3,3-tetramethyl, dichloromethyl dichloromethyl dimethylsilyl oxy dimethylsilane, acmc-20ap5a, disiloxane,3-bis dichloromethyl-1,1,3,3-tetramethyl, disiloxane,1,3-bis dichloromethyl-1,1,3,3-tetramethyl, 1,3-bis dichloromethyl-1,1,3,3-tetramethylpropanedisiloxane
C[Si](C)(C(Cl)Cl)O[Si](C)(C)C(Cl)Cl
300.147
300.16
1.466
1,3-Bis(dichloromethyl)-1,1,3,3-tetramethyldisiloxane
2943-70-6
MFCD00040856
65°C (149°F)
YMDCHYWOAHMASL-UHFFFAOYSA-N
dichloromethyl-[dichloromethyl(dimethyl)silyl]oxy-dimethylsilane
76260
94%
1.221
Videos
Sicherheit und Handhabung

Sicherheit und Handhabung

GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.

P210-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P370+P378q-P501c

H227-H315-H319

missing translation for 'einecsNumber' : 220-939-1

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate