missing translation for 'onlineSavingsMsg'
Learn More
1-(3-Chloro-5-trifluoromethyl-2-pyridyl)homopiperazine, 98%, Thermo Scientific Chemicals Product Code.: 11459400

1-(3-Chloro-5-trifluoromethyl-2-pyridyl)homopiperazine, 98%, Thermo Scientific Chemicals

Product Code. 11459400
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11459400

Brand: Thermo Scientific Alfa Aesar H50263.03

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 231953-40-5
Molecular Formula C11H13ClF3N3
Molecular Weight (g/mol) 279.69
MDL Number MFCD00238856
InChI Key PKKLERQDFWUWNM-UHFFFAOYSA-N
Synonym 1-3-chloro-5-trifluoromethyl pyridin-2-yl-1,4-diazepane, 1-3-chloro-5-trifluoromethyl-2-pyridyl-1,4-diazepane, 1-3-chloro-5-trifluoromethyl-2-pyridyl homopiperazine, 1-3-chloro-5-trifluoromethyl pyrid-2-yl homopiperazine, 1-3-chloranyl-5-trifluoromethyl pyridin-2-yl-1,4-diazepane, acmc-20ap02, 1-3-chloro-5-trifluoromethyl pyridin-2-yl homopiperazine, 1-3-chloro-5-trifluoromethyl-2-pyridinyl-1,4-diazepaneShow MoreShow Less
PubChem CID 735734
IUPAC Name 1-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]-1,4-diazepane
SMILES FC(F)(F)C1=CC(Cl)=C(N=C1)N1CCCNCC1

Specifications

Boiling Point 128°C (1 mmHg)
Quantity 1 g
UN Number UN2735
Sensitivity Air Sensitive
Formula Weight 279.69
Percent Purity 98%
Chemical Name or Material 1-(3-Chloro-5-trifluoromethyl-2-pyridyl)homopiperazine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-(3-Chloro-5-trifluoromethyl-2-pyridyl)homopiperazine, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.