missing translation for 'onlineSavingsMsg'
Learn More

1-(3-Chlorophenoxy)-3-butyn-2-ol, 97%, Thermo Scientific™

Product Code. 11411090
Click to view available options
Quantity:
1 g
5 g
25 g

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86912-84-7
Molecular Formula C10H9ClO2
Molecular Weight (g/mol) 196.63
MDL Number MFCD09909563
InChI Key CEZMXOXWZYDVOU-UHFFFAOYSA-N
Synonym 1-3-chlorophenoxy but-3-yn-2-ol, 1-3-chlorophenoxy-3-butyn-2-ol
PubChem CID 13145644
IUPAC Name 1-(3-chlorophenoxy)but-3-yn-2-ol
SMILES C#CC(COC1=CC(=CC=C1)Cl)O

Specifications

Melting Point 36°C to 40°C
Quantity 1 g
Formula Weight 196.63
Percent Purity 97%
Chemical Name or Material 1-(3-Chlorophenoxy)-3-butyn-2-ol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.