missing translation for 'onlineSavingsMsg'
Learn More

1,3-Cyclohexadiene, 96%, stabilized, Thermo Scientific Chemicals

Product Code. 10487480
Click to view available options
Quantity:
5 mL
25 mL
100 mL
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 592-57-4
Molecular Formula C6H8
Molecular Weight (g/mol) 80.13
MDL Number MFCD00001532
InChI Key MGNZXYYWBUKAII-UHFFFAOYSA-N
Synonym 1,3-cyclohexadiene, cyclohexadiene, 1,2-dihydrobenzene, unii-jv5w0eg5bp, jv5w0eg5bp, 3-cyclohexen-1,2-ylene, 1,3-cyclohexadien, 1,3 cyclohexadiene, 2,4-cyclohexadiene, cyclohexadiene-1,3
PubChem CID 11605
ChEBI CHEBI:37610
IUPAC Name cyclohexa-1,3-diene
SMILES C1CC=CC=C1

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point -98.0°C
Color Colorless to Yellow
Density 0.8600g/mL
Boiling Point 80.0°C
Flash Point −6°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Refractive Index 1.4730 to 1.4760
Quantity 25 mL
Specific Gravity 0.86
Solubility Information Solubility in water: immiscible.
Stabilizer 0.1% BHT
Formula Weight 80.13
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 1,3-Cyclohexadiene, 96%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1,3-Cyclohexadiene, stabilized
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
Supplemental information
  • MIXTURE LIST-Contains : 1,3-Cyclohexadiene

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'