Learn More
1,3-Cyclohexanedione, 97%, Thermo Scientific Chemicals
£40.70 - £433.90
Chemical Identifiers
CAS | 504-02-9 |
---|---|
Molecular Formula | C6H8O2 |
Molecular Weight (g/mol) | 112.13 |
MDL Number | MFCD00001585 |
InChI Key | HJSLFCCWAKVHIW-UHFFFAOYSA-N |
Synonym | 1,3-cyclohexanedione, dihydroresorcinol, 1,3-cyclohexandione, 1,3 cyclohexanedione, hydroresorcinol, resorcinol, dihydro, 1,3-cyclohexanone, unii-6uk3d2bxjt, 1,3-benzenediol, dihydro, 1,3-cyclohexane dione |
PubChem CID | 10434 |
ChEBI | CHEBI:17766 |
IUPAC Name | cyclohexane-1,3-dione |
SMILES | C1CC(=O)CC(=O)C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10112570
|
Thermo Scientific Acros
111171000 |
100 g |
£40.70
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10530021
|
Thermo Scientific Acros
111175000 |
500 g |
£123.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10629833
|
Thermo Scientific Acros
111170025 |
2.5 kg | |||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
504-02-9 | |
112.13 | |
HJSLFCCWAKVHIW-UHFFFAOYSA-N | |
10434 | |
cyclohexane-1,3-dione |
C6H8O2 | |
MFCD00001585 | |
1,3-cyclohexanedione, dihydroresorcinol, 1,3-cyclohexandione, 1,3 cyclohexanedione, hydroresorcinol, resorcinol, dihydro, 1,3-cyclohexanone, unii-6uk3d2bxjt, 1,3-benzenediol, dihydro, 1,3-cyclohexane dione | |
CHEBI:17766 | |
C1CC(=O)CC(=O)C1 |
Specifications
504-02-9 | |
100.0 | |
Beige to Yellow | |
1% max. Sodium chloride (Argentometry) | |
C6H8O2 | |
MFCD00001585 | |
01,178 | |
1,3-cyclohexanedione, dihydroresorcinol, 1,3-cyclohexandione, 1,3 cyclohexanedione, hydroresorcinol, resorcinol, dihydro, 1,3-cyclohexanone, unii-6uk3d2bxjt, 1,3-benzenediol, dihydro, 1,3-cyclohexane dione | |
HJSLFCCWAKVHIW-UHFFFAOYSA-N | |
cyclohexane-1,3-dione | |
10434 | |
112.13 | |
Powder |
96.0 | |
>101°C | |
Authentic | |
Plastic bottle | |
C6H8(=O)2 | |
07, 554 | |
15, 3196 | |
Solubility in water: soluble. Other solubilities: soluble in alcohol,chloroform,acetone and,boiling benzene,slightly soluble in ether,carbon disulfide and,petroleum ether | |
C1CC(=O)CC(=O)C1 | |
112.13 | |
CHEBI:17766 | |
97% | |
1, 3-Cyclohexanedione, 97% |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 207-980-0
missing translation for 'tsca' : TSCA
- 1,3-Cyclohexanedione
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
Hazard Statement
- H302-Harmful if swallowed.
Precautionary Statement
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
RUO – Research Use Only