missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Diacetylbenzene, 97%, Thermo Scientific Chemicals
£21.90 - £54.40
Chemical Identifiers
CAS | 6781-42-6 |
---|---|
Molecular Formula | C10H10O2 |
Molecular Weight (g/mol) | 162.19 |
MDL Number | MFCD00008740 |
InChI Key | VCHOFVSNWYPAEF-UHFFFAOYSA-N |
Synonym | 1,3-diacetylbenzene, 1,1'-1,3-phenylene diethanone, ethanone, 1,1'-1,3-phenylene bis, m-diacetylbenzene, benzene-1,3-bis acetyl, m-acetylacetophenone, m-acetyl acetophenone, 1-3-acetylphenyl ethanone, 1-3-acetylphenyl ethan-1-one, m-diacetyl benzene |
PubChem CID | 23229 |
SMILES | CC(=O)C1=CC(=CC=C1)C(C)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11494937
|
Thermo Scientific Alfa Aesar
B21880.03 |
1 g |
£21.90
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11404947
|
Thermo Scientific Alfa Aesar
B21880.06 |
5 g |
£54.40
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
1,3-Diacetylbenzene was used in the preparation of polyhydroxylated analogs. It is an important raw material and intermediates in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,3-Diacetylbenzene was used in the preparation of polyhydroxylated analogs. It is an important raw material and intermediates in organic synthesis.
Solubility
Soluble in ethanol, benzene, chloroform.
Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.
Chemical Identifiers
6781-42-6 | |
162.19 | |
VCHOFVSNWYPAEF-UHFFFAOYSA-N | |
23229 |
C10H10O2 | |
MFCD00008740 | |
1,3-diacetylbenzene, 1,1'-1,3-phenylene diethanone, ethanone, 1,1'-1,3-phenylene bis, m-diacetylbenzene, benzene-1,3-bis acetyl, m-acetylacetophenone, m-acetyl acetophenone, 1-3-acetylphenyl ethanone, 1-3-acetylphenyl ethan-1-one, m-diacetyl benzene | |
CC(=O)C1=CC(=CC=C1)C(C)=O |
Specifications
6781-42-6 | |
150°C to 155°C (15 mmHg) | |
C10H10O2 | |
2042511 | |
Soluble in ethanol,benzene,chloroform. | |
CC(=O)C1=CC(=CC=C1)C(C)=O | |
162.19 | |
162.19 | |
1,3-Diacetylbenzene |
35°C to 37°C | |
>110°C (230°F) | |
MFCD00008740 | |
1,3-diacetylbenzene, 1,1'-1,3-phenylene diethanone, ethanone, 1,1'-1,3-phenylene bis, m-diacetylbenzene, benzene-1,3-bis acetyl, m-acetylacetophenone, m-acetyl acetophenone, 1-3-acetylphenyl ethanone, 1-3-acetylphenyl ethan-1-one, m-diacetyl benzene | |
VCHOFVSNWYPAEF-UHFFFAOYSA-N | |
1-(3-acetylphenyl)ethan-1-one | |
23229 | |
97% |
Safety and Handling
missing translation for 'einecsNumber' : 229-842-9
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only