missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Diazaspiro[4.5]decane-2,4-dione, 97%, Thermo Scientific™
£17.35 - £52.95
Chemical Identifiers
CAS | 702-62-5 |
---|---|
Molecular Formula | C8H12N2O2 |
Molecular Weight (g/mol) | 168.196 |
MDL Number | MFCD00022404 |
InChI Key | NERNEXMEYQFFHU-UHFFFAOYSA-N |
Synonym | 1,3-diazaspiro 4.5 decane-2,4-dione, 5,5-pentamethylenehydantoin, hydantoin, pentamethylene, 5,5'-cyclohexanespirohydantoin, 5,5-pentamethylenespirohydantoin, hydantoin, 5,5-pentamethylene, cyclohexanespiro-5'-hydantoin, spiro cyclohexane-1,5'-hydantoin, 2,4-diazaspiro 4.5 decane-1,3-dione, cyclohexane-spiro-5-hydantoin |
PubChem CID | 12799 |
IUPAC Name | 1,3-diazaspiro[4.5]decane-2,4-dione |
SMILES | C1CCC2(CC1)C(=O)NC(=O)N2 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10593042
|
Thermo Scientific Maybridge
JFD02410DA |
1 g |
£17.35
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10722357
|
Thermo Scientific Maybridge
JFD02410DE |
5 g |
£52.95
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
702-62-5 | |
168.196 | |
NERNEXMEYQFFHU-UHFFFAOYSA-N | |
12799 | |
C1CCC2(CC1)C(=O)NC(=O)N2 |
C8H12N2O2 | |
MFCD00022404 | |
1,3-diazaspiro 4.5 decane-2,4-dione, 5,5-pentamethylenehydantoin, hydantoin, pentamethylene, 5,5'-cyclohexanespirohydantoin, 5,5-pentamethylenespirohydantoin, hydantoin, 5,5-pentamethylene, cyclohexanespiro-5'-hydantoin, spiro cyclohexane-1,5'-hydantoin, 2,4-diazaspiro 4.5 decane-1,3-dione, cyclohexane-spiro-5-hydantoin | |
1,3-diazaspiro[4.5]decane-2,4-dione |
Specifications
702-62-5 | |
97% | |
C8H12N2O2 | |
1,3-diazaspiro 4.5 decane-2,4-dione, 5,5-pentamethylenehydantoin, hydantoin, pentamethylene, 5,5'-cyclohexanespirohydantoin, 5,5-pentamethylenespirohydantoin, hydantoin, 5,5-pentamethylene, cyclohexanespiro-5'-hydantoin, spiro cyclohexane-1,5'-hydantoin, 2,4-diazaspiro 4.5 decane-1,3-dione, cyclohexane-spiro-5-hydantoin | |
C1CCC2(CC1)C(=O)NC(=O)N2 | |
168.196 | |
168.19 | |
1, 3-Diazaspiro[4.5]decane-2, 4-dione |
97.0 | |
Amber glass bottle | |
MFCD00022404 | |
NERNEXMEYQFFHU-UHFFFAOYSA-N | |
1,3-diazaspiro[4.5]decane-2,4-dione | |
12799 | |
97% |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning