missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Dibromo-2,2-dimethoxypropane, 99%, Thermo Scientific™
£11.10 - £65.10
Chemical Identifiers
CAS | 22094-18-4 |
---|---|
Molecular Formula | C5H10Br2O2 |
Molecular Weight (g/mol) | 261.94 |
MDL Number | MFCD00051792 |
InChI Key | CPAHOXOBYHMHDT-UHFFFAOYSA-N |
Synonym | 1,3-dibromoacetone dimethyl acetal, 1,3-acetone dibromide dimethyl acetal, 1,3-dibromo-2,2-dimethoxy-propane, 2-bromo-1-bromomethyl-1,1-dimethoxyethane, 1,3-dibromoacetone dimethyl ketal, propane, 1,3-dibromo-2,2-dimethoxy, pubchem2043, ag120 intermediates, acmc-1ck33, timtec-bb sbb039320 |
PubChem CID | 2734196 |
IUPAC Name | 1,3-dibromo-2,2-dimethoxypropane |
SMILES | COC(CBr)(CBr)OC |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10436242
|
Thermo Scientific Acros
355090010 |
1 g | Glass bottle | ||||||
10570902
|
Thermo Scientific Acros
355090050 |
5 g | Glass bottle |
£65.10
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
22094-18-4 | |
261.94 | |
CPAHOXOBYHMHDT-UHFFFAOYSA-N | |
2734196 | |
COC(CBr)(CBr)OC |
C5H10Br2O2 | |
MFCD00051792 | |
1,3-dibromoacetone dimethyl acetal, 1,3-acetone dibromide dimethyl acetal, 1,3-dibromo-2,2-dimethoxy-propane, 2-bromo-1-bromomethyl-1,1-dimethoxyethane, 1,3-dibromoacetone dimethyl ketal, propane, 1,3-dibromo-2,2-dimethoxy, pubchem2043, ag120 intermediates, acmc-1ck33, timtec-bb sbb039320 | |
1,3-dibromo-2,2-dimethoxypropane |
Specifications
22094-18-4 | |
100.0 | |
White | |
98.5% min. (GC) | |
MFCD00051792 | |
CPAHOXOBYHMHDT-UHFFFAOYSA-N | |
1,3-dibromo-2,2-dimethoxypropane | |
2734196 | |
99% | |
1, 3-Dibromo-2, 2-dimethoxypropane |
98.5 | |
65°C to 68°C | |
Authentic | |
C5H10Br2O2 | |
1,3-dibromoacetone dimethyl acetal, 1,3-acetone dibromide dimethyl acetal, 1,3-dibromo-2,2-dimethoxy-propane, 2-bromo-1-bromomethyl-1,1-dimethoxyethane, 1,3-dibromoacetone dimethyl ketal, propane, 1,3-dibromo-2,2-dimethoxy, pubchem2043, ag120 intermediates, acmc-1ck33, timtec-bb sbb039320 | |
COC(CBr)(CBr)OC | |
261.94 | |
261.94 | |
Crystalline Powder |
Safety and Handling
missing translation for 'tsca' : TSCA
RUO – Research Use Only